Learn More
DWK Life Sciences DURAN™ Standard Cone, diamond-ground
Designed for a rigid, self-supporting connection with lower requirements.
151.00 € - 229.00 €
Specifications
Quantity 50/Pk.
View More Specs
Products 4
Product Code Brand Joint Size Price Quantity & Availability  
Product Code Brand Joint Size Price Quantity & Availability  
15884558
View Documents
DWK Life Sciences
286154208
13 mm
151.00 €
Pack of 50
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
15981531
View Documents
DWK Life Sciences
286158801
26 mm
229.00 €
Pack of 50
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
15854568
View Documents
DWK Life Sciences
286156303
22 mm
205.00 €
Pack of 50
In Stock
to see stock.
Add to basket
15824568
View Documents
DWK Life Sciences
286155401
17 mm
194.00 €
Pack of 50
In Stock
to see stock.
Add to basket
Description
Description
The conical ground joint of sleeve and core is a rigid, self-supporting joint. DURAN™ DG cores are not ground according to DIN 12249 and represent the cost-effective alternative to DURAN™ cores. Ground joints with ground grease or PTFE sleeves should always be used. KECK™ Conical ground joint clamps are the perfect supplement for securing the conical ground joint.
  • Diamond cut (coarser cut quality, more roughness depth)
  • Cost-effective alternative to DURAN™ cores (not according to DIN)
  • Tight closure due to standard ground joint
  • Glass articles for further processing
Specifications
Specifications
50/Pk.