Learn More
DWK Life Sciences DURAN™ Standard Cone with tip, finished
Core cut with extension.
Brand:  DWK Life Sciences 286165205
 View more versions of this product
Product Code. 15844588
  • 201.00 € / Pack of 50
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The DURAN™ cores with outlet comply with DIN 12249 Form G. The process has the function of a drip ring. It prevents the liquid from running down the inner wall of the vessel. For this purpose, the outlet is usually cut at an angle and fused. At the same time, mediums do not come into contact with the greased ground surface. The additional outlet can also be used as an attachment point for extensions for various purposes, e.g. as an inlet pipe.
  • Ground (finer grinding quality, less roughness depth)
  • The additional drain on the grinding enables a clean drain/leakage of liquids without wetting the grinding.
  • Tight closure due to standard ground joint
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50/Pk.
The additional outlet can also be used as an attachment point for extensions for various purposes, e.g. as an inlet pipe; Installation in glass apparatus and devices
17 mm
1.8 mm