Learn More
DWK Life Sciences DURAN™ Standard Cone with tip, finished
Core cut with extension.
3.71 € - 518.00 €
Products 16
Product Code Brand Joint Size Price Quantity & Availability  
Product Code Brand Joint Size Price Quantity & Availability  
15911541
View Documents
DWK Life Sciences
286166801
26 mm
257.00 €
Pack of 50
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
15824588
View Documents
DWK Life Sciences
286164406
13 mm
166.00 €
Pack of 50
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
15844588
View Documents
DWK Life Sciences
286165205
17 mm
201.00 €
Pack of 50
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
15814588
View Documents
DWK Life Sciences
286164003
11 mm
205.00 €
Pack of 50
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
15874588
View Documents
DWK Life Sciences
286168605
30 mm
91.65 €
Pack of 10
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
15884588
View Documents
DWK Life Sciences
286169404
40 mm
126.00 €
Pack of 10
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
15804598
View Documents
DWK Life Sciences
286170808
46 mm
203.00 €
Pack of 10
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
15894578
View Documents
DWK Life Sciences
286163007
6 mm
74.65 €
Pack of 10
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
10182121
View Documents
DWK Life Sciences
286166004
22 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10429732
View Documents
DWK Life Sciences
286169601
90 mm
111.00 €
Each
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
15804588
View Documents
DWK Life Sciences
286163401
8 mm
241.00 €
Pack of 50
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
15894588
View Documents
DWK Life Sciences
286170208
36 mm
126.00 €
Pack of 10
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
15814598
View Documents
DWK Life Sciences
286171007
50 mm
234.00 €
Pack of 10
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
15824598
View Documents
DWK Life Sciences
286171204
56 mm
276.00 €
Pack of 10
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
15834598
View Documents
DWK Life Sciences
286171607
65 mm
518.00 €
Pack of 10
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
10489152
View Documents
DWK Life Sciences
286172209
80 mm
79.55 €
Each
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
Description
Description
The DURAN™ cores with outlet comply with DIN 12249 Form G. The process has the function of a drip ring. It prevents the liquid from running down the inner wall of the vessel. For this purpose, the outlet is usually cut at an angle and fused. At the same time, mediums do not come into contact with the greased ground surface. The additional outlet can also be used as an attachment point for extensions for various purposes, e.g. as an inlet pipe.
  • Ground (finer grinding quality, less roughness depth)
  • The additional drain on the grinding enables a clean drain/leakage of liquids without wetting the grinding.
  • Tight closure due to standard ground joint
  • Glass articles for further processing