Learn More
DWK Life Sciences DURAN™ Tube with two Hose Connections
For attaching to a wide variety of apparatus.
Brand:  DWK Life Sciences 286021407
304.95 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15864508
  • 354.00 € / Pack of 500
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Hoses can be connected to laboratory apparatus via DURAN™ tube nozzles. There is always a risk of injury due to glass breakage when dismantling tightly fitted hoses from glass olives. DWK therefore recommends the use of DURAN™ glass threads with plastic nozzles.
  • Hose olives enable the connection of hoses to laboratory apparatuses
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
500/Pk.
For attaching to various apparatuses; Installation in different apparatus possible
8 mm