Learn More
DWK Life Sciences DURAN™ Tube with two Hose Connections
For attaching to a wide variety of apparatus.
168.00 € - 354.00 €
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
15854508
|
DWK Life Sciences
286021004 |
6 mm |
168.00 €
Pack of 250 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
15864508
|
DWK Life Sciences
286021407 |
8 mm |
354.00 €
Pack of 500 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
15874508
|
DWK Life Sciences
286021604 |
7 mm |
354.00 €
Pack of 500 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
15961501
|
DWK Life Sciences
286022206 |
9 mm |
178.00 €
Pack of 250 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
15894508
|
DWK Life Sciences
286023202 |
11 mm |
229.00 €
Pack of 250 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
15884508
|
DWK Life Sciences
286022403 |
10 mm |
229.00 €
Pack of 250 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
15804518
|
DWK Life Sciences
286024404 |
14 mm |
173.00 €
Pack of 125 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
15252109
|
DWK Life Sciences
286023605 |
12 mm |
244.00 €
Pack of 250 |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
Description
Hoses can be connected to laboratory apparatus via DURAN™ tube nozzles. There is always a risk of injury due to glass breakage when dismantling tightly fitted hoses from glass olives. DWK therefore recommends the use of DURAN™ glass threads with plastic nozzles.
- Hose olives enable the connection of hoses to laboratory apparatuses
- Glass articles for further processing