missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DyNAmo HS SYBR Green qPCR Kit

Contains a modified Thermus brockianus DNA polymerase and all the other reagents needed for qPCR.

Brand:  Thermo Scientific™ F410XL

 View more versions of this product

Product Code. 10206079

  • 2815.00 € / 2500 reactions

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Only template DNA and PCR primers need to be added by the user. The modified DNA polymerase in this kit incorporates a non-specific DNA-binding domain that lends physical stability to the polymerase-DNA complex. ROX passive reference dye is also supplied with the kit in a separate tube for instruments that require ROX normalization.

  • Strong specificity: DyNAmo HS SYBR Green master mix minimizes the formation of unwanted primer-dimers and extension of non-specifically bound primers during reaction setup
  • Highly sensitive detection: Optimized master mix allows reproducible detection of low-copy number templates
  • Broad dynamic range: DyNAmo HS SYBR Green qPCR Kit detects as few as one copy of the target gene and achieves a dynamic range of up to eight orders of magnitude
  • Convenient to use: 2X master mix, one tube only; ROX passive reference dye provided in a separate vial
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contains:
• 25 x1 mL DyNAmo HS SYBR Green qPCR Mix (2X)
• 1 x 1.25 mL 50 x ROX dye, 25μM

Store at -20°C
Tube
qPCR
Standard
DyNAmo HS SYBR Green qPCR Kit
Real Time PCR SYBR Master Mix
2500 Reactions of 20 μL Each, 1000 Reactions of 50 μL Each
2X (DyNAmo HS SYBR Green qPCR Mix), 50X (ROX Dye)
DNA, dsDNA
SYBR
High
Tbr DNA polymerase
7000 System, 7300 System, 7500 Fast System, 7500 System, 7700 System, 7900HT Fast System, 7900HT System, BioRad CFX96™, QuantStudio™, ViiA™ 7 System, Bio-Rad CFX384 Touch™, Roche LightCycler 480, 5700 Sequence Detection System, StepOnePlus™ System, Eppendorf MasterCycler RealPlex, Corbett RotorGene, StepOne™ System, Stratagene Mx4000, Stratagene Mx3005P, Stratagene Mx3000P, MJ Opticon, MJ Chromo4, Cepheid SmartCycler
DyNAmo
2,500 reactions
Gene Expression
2500 Reactions
25 x 1 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.