Learn More
Dysprosium(III) oxalate decahydrate, REacton™, 99.9% (REO), Thermo Scientific™
54.20 € - 162.00 €
Chemical Identifiers
CAS | 24670-07-3 |
---|---|
Molecular Formula | C6H26Dy2O22 |
Molecular Weight (g/mol) | 775.252 |
MDL Number | MFCD00150482 |
InChI Key | WJYBVSNZIQHDSA-UHFFFAOYSA-N |
Synonym | scandium oxalate decahydrate |
PubChem CID | 131876564 |
IUPAC Name | dysprosium;oxalic acid;decahydrate |
SMILES | C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.O.O.O.O.O.O.O.O.O.O.[Dy].[Dy] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
24670-07-3 | |
775.252 | |
WJYBVSNZIQHDSA-UHFFFAOYSA-N | |
131876564 | |
C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.O.O.O.O.O.O.O.O.O.O.[Dy].[Dy]Show More |
C6H26Dy2O22 | |
MFCD00150482 | |
scandium oxalate decahydrate | |
dysprosium;oxalic acid;decahydrate |
Specifications
40°C (in -H2O) | |
White | |
C6H26Dy2O22 | |
MFCD00150482 | |
scandium oxalate decahydrate | |
C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.O.O.O.O.O.O.O.O.O.O.[Dy].[Dy] | |
775.252 | |
769.21 (589.06 Anhydrous) | |
Reacton™ | |
(REO) |
24670-07-3 | |
Powder | |
Dy2(C2O4)3·10H2O | |
UN3288 | |
WJYBVSNZIQHDSA-UHFFFAOYSA-N | |
dysprosium;oxalic acid;decahydrate | |
131876564 | |
99.9% | |
Hygroscopic | |
Dysprosium(III) oxalate decahydrate |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.