Learn More
Invitrogen™ E-Gel™ Agarose Gels with SYBR™ Safe DNA Gel Stain
GREENER_CHOICE
Invitrogen E-Gel precast agarose gels with SYBR Safe DNA gel stain are designed for fast, convenient, and safe DNA sample electrophoresis.
Brand:  Invitrogen™ A45204
 View more versions of this product
Product Code. 16438869
  • 247.00 € / Pack of 2
Explore more special offers
Description
Show More
Specifications
Specifications
E-Gel™ 1 Kb Plus Express Ladder
2%
Agarose Gel
SYBR™ Safe
Less hazardous
Room Temperature
Each box contains 10 gels
  • Store strictly at room temperature.
  • Do not freeze.
  • Avoid cold spots near freezers or other uncontrolled environments.
E-Gel
50 to 2000 bp
11-well
2 x 10 Gels
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ E-Gel™ Agarose Gels with SYBR™ Safe DNA Gel Stain > Agarose Gels with SYBR™ Safe DNA Gel Stain, 2%, 2 x 10 gels