Learn More
Berkshire™ EasyClean™ 360 System

360 System includes a light-weight, maneuverable cleaning tool along with easy-to-use, laundered polyester knit covers.
Brand: Berkshire™ EC360CHPST40
Description
360 System includes a light-weight, maneuverable cleaning tool along with easy-to-use, laundered polyester knit covers.
- Sterile, 100% continuous filament polyester knit covers
- Cleanroom laundered, knife cut edge
- Certificate of Sterility with each case and available on the web by lot number 24/7*
*Details: Expiration Date, Radiation Dosage, Lot Information, Meets cGMP Requirements For Traceability.
Specifications
Choice Isolator Tool Cover | |
Autoclavable | |
Polyester Knit (1%) |
Sterile | |
No |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.