Learn More
Ebro Electronic™ EBI 20 Data Logger Series
Data logger to measure temperature and / or humidity during transportation and in warehouses
Brand:  Ebro Electronic™ 1601-0043A
 View more versions of this product
Product Code. 10493150
  • 140.00 € / Each
Explore more special offers
Description
Description
EBI 20 data loggers are very robust and are in use all over the world in transportation and for warehouse monitoring.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Datalogger
40,000 Readings
Flashing LCD Segment
>24 Months
Factory Calibration certificate
EBI 20-TE1
Reliable Temperature Recording, Core Temperature Measurement, Annual memory, Transport, Storage monitoring, Refrigerating and deep-freeze rooms, Refrigerated display cases
0.1°C
69 x 48 x 22 mm
45 g
LCD
Calibrated
Lithium
1
IP67
NTC
±0.5 at –20 to 40 °C
-30°C to +60°C
69 mm