missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elma™ EC110 Concentrated Cleaner
Providing powerful cleaning power for labware and medical instruments. Elma™ EC110 Concentrated Cleaner is an alkaline ultrasonic solution.
Brand: Elma™ 5807900000
This item is not returnable.
View return policy
Description
- Suitable for metal, ceramic and quarz surfaces, surfaces of numerous plastic materials, rubber surfaces and particularly for the cleaning of silicium wafers after mechanical treatment (sawing)
- Not suitable for light metals (aluminum, magnesium and their alloys) and plastics that are susceptible for stress cracking
- Removes metal oxides, stains of quarz and tenacious organic contaminants such as scaled and gummy oil and grease, wax, residual lacquer and paint
Specifications
EC 110 Cleaner | |
12.4 | |
>100°C | |
Anionic surfactants <5%, non-ionic surfactants <5%, Phosphonates <5% | |
Can | |
For cleaning Metal, Ceramic, Plastics, Silicium wafer and Quartz-surfaces |
Liquid | |
Brown | |
Annex VII A, EC 648/2004, UN 1824 | |
Ultrasonic bath 3 to 5% | |
5°C to 30°C | |
25 L |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- elma clean 110 (EC 110)
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H400-Very toxic to aquatic life.
Precautionary Statement
- P102-Keep out of reach of children.
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P405-Store locked up.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction