missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elma™ EC110 Concentrated Cleaner
Providing powerful cleaning power for labware and medical instruments. Elma™ EC110 Concentrated Cleaner is an alkaline ultrasonic solution.
21.55 € - 224.00 €
Specifications
Product Type | EC 110 Cleaner |
---|---|
Form | Liquid |
pH | 12.4 |
Appearance | Brown |
Boiling Point | >100°C |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
11791359
|
Elma™
5807900000 |
25 L |
224.00 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12566319
|
Elma™
5808500000 |
1 L |
21.55 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Suitable for metal, ceramic and quarz surfaces, surfaces of numerous plastic materials, rubber surfaces and particularly for the cleaning of silicium wafers after mechanical treatment (sawing)
- Not suitable for light metals (aluminum, magnesium and their alloys) and plastics that are susceptible for stress cracking
- Removes metal oxides, stains of quarz and tenacious organic contaminants such as scaled and gummy oil and grease, wax, residual lacquer and paint
Specifications
EC 110 Cleaner | |
12.4 | |
>100°C | |
Ultrasonic bath 3 to 5% | |
For cleaning Metal, Ceramic, Plastics, Silicium wafer and Quartz-surfaces |
Liquid | |
Brown | |
Annex VII A, EC 648/2004, UN 1824 | |
5°C to 30°C |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- elma clean 110 (EC 110)
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H400-Very toxic to aquatic life.
Precautionary Statement
- P102-Keep out of reach of children.
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P405-Store locked up.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title