missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elma™ EC110 Concentrated Cleaner
Providing powerful cleaning power for labware and medical instruments. Elma™ EC110 Concentrated Cleaner is an alkaline ultrasonic solution.
Brand: Elma™ 5808500000
This item is not returnable.
View return policy
Description
- Suitable for metal, ceramic and quarz surfaces, surfaces of numerous plastic materials, rubber surfaces and particularly for the cleaning of silicium wafers after mechanical treatment (sawing)
- Not suitable for light metals (aluminum, magnesium and their alloys) and plastics that are susceptible for stress cracking
- Removes metal oxides, stains of quarz and tenacious organic contaminants such as scaled and gummy oil and grease, wax, residual lacquer and paint
Specifications
EC 110 Cleaner | |
12.4 | |
>100°C | |
Ultrasonic bath 3 to 5% | |
For cleaning Metal, Ceramic, Plastics, Silicium wafer and Quartz-surfaces |
Liquid | |
Brown | |
Annex VII A, EC 648/2004, UN 1824 | |
5°C to 30°C | |
1 L |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P102-Keep out of reach of children.
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : alkylpolyglycoside