Learn More
Macherey-Nagel™ EC Analytical HPLC Column
Brand:  Macherey-Nagel™ 720046.40
 View more versions of this product
Product Code. 10002951
  • 1450.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Specifications
BSA
4 mm
For HPLC, GC,TLC, SPE Application
5800 psi (400 bar)
Spherical
5 to 8
100 m²/g
L75
Silica
No
Analytical EC Column
Analytical Column
0.7 mL/min
150 mm
50 mmol/L/L phosphate buffer + 1 % 1-propanol
7 μm
300 Å
40°C
BSA-7
Mixed Mode
HIC, Polar Selectivity, Steric Selectivity
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ EC Analytical HPLC Column > Stationary Phase: BSA-7