missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ EC Analytical HPLC Column

1085.00 € - 1450.00 €

Specifications

Column Format Analytical Column
Diameter (Metric) 4 mm
Packing Material Silica
Type Analytical EC Column
View More Specs

Products 5
Product Code Brand Stationary Phase Price Quantity & Availability  
Product Code Brand Stationary Phase Price Quantity & Availability  
10002951
View Documents
Macherey-Nagel™
720046.40
BSA-7
1450.00 €
Each
In Stock
to see stock.
Add to basket
 
10788612
View Documents
Macherey-Nagel™
720127.40
Permethylated α-cyclodextrin
1295.00 €
Each
In Stock
to see stock.
Add to basket
 
10556141
View Documents
Macherey-Nagel™
720124.40
β-cyclodextrin
1085.00 €
Each
In Stock
to see stock.
Add to basket
 
10105131
View Documents
Macherey-Nagel™
720125.40
Permethylated β-cyclodextrin
1295.00 €
Each
In Stock
to see stock.
Add to basket
 
10706512
View Documents
Macherey-Nagel™
720752.40
Permethylated γ-cyclodextrin
1430.00 €
Each
Estimated Shipment: 25-02-2025
to see stock.
Add to basket
 
Description

Description

Specifications

Specifications

Analytical Column
4 mm
Silica
Analytical EC Column
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.