missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sharpsguard™ Eco 22 Waste Container

The new Sharpsguard™ eco 22 container, made using at least 40% recycled plastic that is used for the disposal of sharps that have not been contaminated with medicines.
31.30 €
Specifications
Capacity (Metric) | 17.86 L |
---|---|
Diameter (Metric) Outer | 327 mm |
Height (Metric) | 343 mm |
Quantity | 1 Ea. |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
17734854
![]() |
Sharpsguard™
DD475OLR |
Orange |
31.30 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17744854
![]() |
Sharpsguard™
DD475YLR |
Yellow |
31.30 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Sharpsguard™ eco 22 is ideal for theatres, laboratories and sluice rooms with high volumes of hazardous waste. The wide aperture allows economical and convenient disposal of large items.
- This container is supplied with the base and lid separately. Details about how to lock the container and keep the users and contents safe can be found on the label.
- It is designed to fit safely and securely into our POUDS™ mobile holders and bracket range.
- This container is for disposal by incineration or other authorized process and is designed for the consignment of waste categorized by EWC codes: 18-01-01, 18-01-03, 18-02-01, 18-02-02.
Specifications
17.86 L | |
327 mm | |
343 mm | |
1 Ea. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sharpsguard™ Eco 22 Waste Container