Learn More
EcoLume LC Fluid, MP Biomedicals
Liquid Scintillation Cocktails; EcoLume; MP Biomedical; Biodegradable
Brand:  MP Biomedicals 0188247004
 View more versions of this product
Product Code. 15732252
  • 875.00 € / 15.14 L
Explore more special offers
Specifications
Liquid Scintillation Cocktails
4 x 1 gal.
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • ecolume ES
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Skin sensitiser Category 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : linear alkyl benzene, nonylphenol, ethoxylated, phenylxylyleythane-PXE