missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EcoLume LC Fluid, MP Biomedicals
Liquid Scintillation Cocktails; EcoLume; MP Biomedical; Biodegradable
307.00 € - 875.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15722252
|
MP Biomedicals
0188247001 |
1 gal. |
307.00 €
3.79 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15732252
|
MP Biomedicals
0188247004 |
4 x 1 gal. |
875.00 €
15.14 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specifications
Liquid Scintillation Cocktails |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- EcoLume Liquid Scintillation Fluid
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethoxylated nonylphenol, phenylxylyethane-PXE