Learn More
Hanna Instruments™ Electrolyte Fill Solution
Contains 3.5M KCl for double junction electrodes and 3.5M + AgCl for single junction. Hanna Instruments™ Electrolyte Fill Solution ensures optimum measurement performance from a single junction pH or ORP electrode.
Brand:  Hanna Instruments™ HI7071
 View more versions of this product
Product Code. 11793639
  • 126.00 € / Pack of 4
Estimated Shipment: 13-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
pH and ORP electrode fill solution that contains 3.5M KCl which is specifically made for double junction electrodes and 3.5M KCl + AgCl specifically for single junction electrodes
  • Convenient 30 mL bottles
  • Clearly marked bottle with expiration date and lot number
  • Four bottles of 30 mL 3.5M KCl (+ AgCl for single junction) electrolyte solution that ensures optimum measurement performance from your single junction pH or ORP electrode.
Specifications
Specifications
30 mL
Electrolyte Fill Solution
Single
3.5M KCl + AgCl
Electrolyte
4 Pack