Learn More
Hanna Instruments™ Electrolyte Fill Solution
Contains 3.5M KCl for double junction electrodes and 3.5M + AgCl for single junction. Hanna Instruments™ Electrolyte Fill Solution ensures optimum measurement performance from a single junction pH or ORP electrode.
126.00 €
Specifications
Capacity (Metric) | 30 mL |
---|---|
Type | Electrolyte Fill Solution |
Quantity | 4 Pack |
Description
pH and ORP electrode fill solution that contains 3.5M KCl which is specifically made for double junction electrodes and 3.5M KCl + AgCl specifically for single junction electrodes- Convenient 30 mL bottles
- Clearly marked bottle with expiration date and lot number
- Four bottles of 30 mL 3.5M KCl (+ AgCl for single junction) electrolyte solution that ensures optimum measurement performance from your single junction pH or ORP electrode.
Specifications
30 mL | |
Electrolyte Fill Solution | |
4 Pack |