missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Solarus™ Electronic Burette

Digital, solar-powered display for easy reading of titration results. High accuracy with double seal PTFE compound piston.

Brand:  Hirschmann™ 9392020

 View more versions of this product

Product Code. 10205991

  • 1360.00 € / Each
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For sensitive manual titration
  • Media conveying parts can be steam sterilized at 121°C
  • Free rotation
  • Media recirculating system for simple and rapid venting without reagent loss
  • Solar panels being the power supply for the display
  • Use within a temperature range of 4°C to 50°C
  • Individual quality certificate with conformity certificate
Specifications

Specifications

Autoclavable
Glass
±0.20 mL
Class A
20 mL
Environmental Fieldwork, Food or Beverage Analysis, General Chemistry, Industrial Titrations, Wastewater Treatment Applications
Electronic Buret
ASTM E287, DIN EN ISO 385
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers