missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hirschmann™ Solarus™ Electronic Burette
Digital, solar-powered display for easy reading of titration results. High accuracy with double seal PTFE compound piston.
1340.00 € - 1360.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10205991
|
Hirschmann™
9392020 |
20 mL |
1360.00 €
Each |
Estimated Shipment: 06-02-2025
Log in to see stock. |
|||||
11632249
|
Hirschmann™
9392010 |
10 mL |
1360.00 €
Each |
In Stock
Log in to see stock. |
|||||
10093991
|
Hirschmann™
9392050 |
50 mL |
1340.00 €
Each |
Estimated Shipment: 04-02-2025
Log in to see stock. |
|||||
Description
- For sensitive manual titration
- Media conveying parts can be steam sterilized at 121°C
- Free rotation
- Media recirculating system for simple and rapid venting without reagent loss
- Solar panels being the power supply for the display
- Use within a temperature range of 4°C to 50°C
- Individual quality certificate with conformity certificate
Specifications
Autoclavable | |
Electronic Buret |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title