Learn More
Kartell Embedding Rings
Embedding rings made from ABS plastic.
Brand:  Kartell 0291904
 View more versions of this product
Product Code. 18028031
  • 237.00 € / Pack of 2000
Explore more special offers
Description
Description
  • Embedding supports suitable for the processing and the histological inclusion of paraffin pieces
  • The inclusion rings can use the same microtome support (instrument used to make the histological sections of samples) of the inclusion cassettes
  • They can be set up on every kind of microtome thanks to their structure and to the possibility to obtain uniform paraffin blocks.
  • Embedding rings is an easy-filling and, after cutting, is always ready for following sections
Specifications
Specifications
Embedding Ring
Blue
28 mm
1.1 in.
0.43 in.
Plastic
41 mm
1.61 in.
11 mm
Single-use