Learn More
Merck Millicell-ERS2 Volt-Ohm Meter and accessories
Measures Trans Epithelial Electrical Resistance (TEER) of epithelial cells in culture
Brand:  Merck MERSSTX00
 View more versions of this product
Product Code. 11740363
  • 5230.00 € / Each
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
Includes: Millicell-ERS2 comes with meter, battery, STX01 electrode, 1,000 Ω test electrode, battery charger. Specialized electrodes for Millicell-96 well plate are available (Cat. No. MERSSTX00).
Description
Description
Due to the small size of the electrodes, the user can easily measure transepithelial voltage and the resistance of cells grown on microporous membranes.
Services
Electrodes and batteries are warranted for 30 days only from the date on which the customer receives these items
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Specialized Electrode
19 cm
2.3 in.
12.5 Hz
Specialized electrodes for millicell-96 well plate
8 to 10 hr.
10° to 38°C
1.4 kg
7.3 in.
Millicell-96 well plate specialized electrodes for volt-ohmmeter readings
6 cm
0 to 90%
1 Pair
50° to 100°F
3 lb.
11 cm