missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck PureProteome™ Protein A/G Mix Magnetic Bead System
powerful system to isolate antibodies faster, easier, and with better reproducibly then ever before
545.00 €
Specifications
Product Type | Protein A/G mix Magnetic bead |
---|---|
Particle Size | 10μm |
Content And Storage | 2° to 8°C |
For Use With (Application) | Immunoprecipitation, cross-linking immunoprecipitation, Antibody purification |
Matrix | Magnetic Beads |
Description
The PureProteome Protein A/G Mix Magnetic Bead System provides researchers with the power of the combination of both Protein A and Protein G immunoglobulin binding affinities in one convenient magnetic bead product, resulting in greater antibody compatibility than either Protein A or Protein G alone.
- Suitable for immunoprecipitation, crosslinking immunoprecipitation, antibody purification
- Binding capacity: 2–3 mg of Rabbit IgG per mL of suspension
- High capacity: Advanced chemistry combined with high surface area provides more binding sites for proteins
- Consistent results with no sample loss: Particles visible as they adhere to side of tube for quick and easy aspiration and complete buffer removal
- Fast processing time: Beads are immobilized in seconds; increased kinetics of bead-protein binding enables shorter incubations
- Particle Size: 10μm
Specifications
Protein A/G mix Magnetic bead | |
2° to 8°C | |
Magnetic Beads |
10μm | |
Immunoprecipitation, cross-linking immunoprecipitation, Antibody purification |