Learn More
Edwards Vacuum™ EMF10 Mist Element
For use with the EMF10 Oil Mist Exhaust Filter.
Brand: Edwards Vacuum™ A22304198
Description
- Rated flow of 12 m3h
- Maximum back pressure of 12 psig
- Relief pressure of 20 psig
- Replacement/spare part
- Manufactured from epoxy impregnated borosilicate glass fiber
Specifications
EMF10 Oil Mist Element | |
Epoxy-impregnated Borosilicate Glass Fiber | |
12 oz. |
Used to separate and trap oil mist and odors discharged with the exhaust from oil sealed rotary vacuum pumps. | |
Back pressure of 12 psig, relief pressure of 20 psig |
Product Content Correction
Product Title
Edwards Vacuum™ EMF10 Mist Element > For use with: E-LAB 2 Rotary Pump
Spot an opportunity for improvement?Share a Content Correction