missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ End-Zone™ End Point Detection Starter Kit

End-Zone end point detection eliminates guesswork and ensures sample dryness by alerting when sample moisture levels have reached the user-selected level. For use with FreeZone™ Freeze Dryers with Lyo-Works OS.

Brand:  Labconco™ 780902000

 View more versions of this product

Product Code. 17218960

  • 396.00 € / Each
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Add to basket

Includes: 1 vacuum sensor and 1 Fast-Freeze Flask top with an opening to insert the vacuum sensor

Requires : FreeZone Freeze Dryer, End-Zone Starter Kit, 600ml or larger Fast-Freeze Flask
This item is not returnable. View return policy

Description

Description

  • Compares vacuum level of empty valve to vacuum level inside Fast-Freeze Flask containing sample
  • Operating with FreeZone’s Lyo-Works OS, it provides alert on display screen
  • May be programmed to send email to user when end point is detected
  • FreeZone Freeze Dryer with Lyo-Works OS, drying chamber or manifold with valve ports, vacuum pump, and 600mL or larger Fast-Freeze Flask are required for operation (not included)
  • End-Zone is not compatible with FreeZone Stoppering Tray Dryers, Bulk Tray Dryers or FreeZone Legacy Freeze Dryers
  • For detecting end point in more than one sample (up to five), order both End-Zone Starter Kit and Add-On Kit(s)
  • Starter Kit includes components for detecting end point in one sample flask
  • Each End-Zone Starter Kit includes: two vacuum sensors, one valve adapter for connecting the reference sensor to an empty port, one Fast-Freeze Flask top with sensor port, connection cable, and mountable holder with space for six sensors
  • Holder may be mounted directly onto freeze dryer
  • Add-On Kit includes components for detecting end point in one sample flask
  • Each End-Zone Add-On Kit includes: one vacuum sensor and one Fast-Freeze Flask top with opening to insert vacuum sensor
  • Requires End-Zone Starter Kit
  • End-Zone provides alternative to visual inspection making it easy to determine when freeze drying is complete
  • End-Zone will alert when sample moisture levels have reached user-selected level

Services

One year

Specifications

Specifications

End Point Detection Accessory
2.9 in.
2.9 in.
9.1 in.
1 vacuum sensor and 1 Fast-Freeze Flask top with an opening to insert the vacuum sensor
7.3 cm
7.3 cm
23.1 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.