Learn More
Memmert™ Entry Port 14 mm for UF75
Optional entry port for Memmert Ovens.
Brand:  Memmert™ UF75D6
Product Code. 17559655
  • 221.00 € / Each
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Entry port with 14 mm diameter
  • Flap closure
  • Custom position TBC in the back
Specifications
Specifications
UF75 Oven
Entry Port
14 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Memmert™ Entry Port 14 mm for UF75 > For Use With: UF75