Learn More
Memmert™ Entry Port Left Centre/Centre
For introducing connections at the side. Moisture tight and can be closed by flap and silicone stopper
138.00 € - 240.00 €
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
17109834
|
Memmert™
HPP410ecoF0 |
HPP410eco |
240.00 €
Each |
Estimated Shipment: 27-02-2025
Log in to see stock. |
|||||
17119694
|
Memmert™
HPP110ecoF0 |
HPP110eco |
240.00 €
Each |
Estimated Shipment: 27-02-2025
Log in to see stock. |
|||||
17130065
|
Memmert™
IPP1060ecoF0 |
IPP1060eco |
138.00 €
Each |
Estimated Shipment: 27-02-2025
Log in to see stock. |
|||||
17140105
|
Memmert™
IPP110ecoF0 |
IPP110eco |
138.00 €
Each |
Estimated Shipment: 27-02-2025
Log in to see stock. |
|||||
17159644
|
Memmert™
HPP1060ecoF0 |
HPP1060eco |
240.00 €
Each |
Estimated Shipment: 27-02-2025
Log in to see stock. |
|||||
17159784
|
Memmert™
HPP260ecoF0 |
HPP260eco |
240.00 €
Each |
Estimated Shipment: 27-02-2025
Log in to see stock. |
|||||
17169874
|
Memmert™
HPP750ecoF0 |
HPP750eco |
240.00 €
Each |
Estimated Shipment: 27-02-2025
Log in to see stock. |
|||||
17170225
|
Memmert™
IPP410ecoF0 |
IPP410eco |
138.00 €
Each |
Estimated Shipment: 27-02-2025
Log in to see stock. |
|||||
17180185
|
Memmert™
IPP260ecoF0 |
IPP260eco |
138.00 €
Each |
Estimated Shipment: 27-02-2025
Log in to see stock. |
|||||
17190265
|
Memmert™
IPP750ecoF0 |
IPP750eco |
138.00 €
Each |
Estimated Shipment: 27-02-2025
Log in to see stock. |
|||||
Description
- Standard position left centre/centre
- Not for model size 260 with light module
- Not for model size 110 with light module
Specifications
Moisture tight |