missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPLIN Mouse anti-Human, Rat, Clone: 6E6E6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66071-1-IG-20UL
This item is not returnable.
View return policy
Description
This antibody recognizes both of isoforms of EPLN.
EPLIN is a cytoskeleton-associated protein that inhibits actin filament depolymerization and cross-links filaments in bundles.Specifications
EPLIN | |
Monoclonal | |
0.55 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9UHB6 | |
LIMA1 | |
EPLIN Fusion Protein Ag13288 | |
20 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry | |
6E6E6 | |
Unconjugated | |
LIMA1 | |
EPLIN, LIM domain and actin binding 1, LIMA1, SREBP3 | |
Mouse | |
Protein G | |
RUO | |
300228, 51474 | |
-20°C | |
Liquid |