missing translation for 'onlineSavingsMsg'
Learn More

EPLIN Mouse anti-Human, Rat, Clone: 6E6E6, Proteintech

Mouse Monoclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen EPLIN
Clone 6E6E6
Concentration 0.55 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16837615
missing translation for 'viewDocuments'
Proteintech
66071-1-IG-20UL
20 μL
145.00 €
20µL
Please to purchase this item. Need a web account? Register with us today!
16827615
missing translation for 'viewDocuments'
Proteintech
66071-1-IG-150UL
150 μL
410.00 €
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody recognizes both of isoforms of EPLN.

EPLIN is a cytoskeleton-associated protein that inhibits actin filament depolymerization and cross-links filaments in bundles.
Specifications

Specifications

EPLIN
0.55 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
EPLIN, LIM domain and actin binding 1, LIMA1, SREBP3
LIMA1
IgG1
Protein G
Antibody
6E6E6
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry
Unconjugated
Mouse
Human, Rat
Q9UHB6
300228, 51474
EPLIN Fusion Protein Ag13288
Primary
-20°C
LIMA1
Videos
SDS
Documents

Documents

Certificates