missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf epMotion™ 96 Flex
Elevate your lab workflows with the epMotion™ 96 Flex. Process 96- and 384-well plates efficiently and benefit from ISO-calibrated and exchangeable dispensing heads. The ergonomic operation is also simple in laminar flow cabinets. Please order the appropriate dispensing head and power cable separately.
Brand: Eppendorf 5069000519
This item is not returnable.
View return policy
Description
- Designed for simultaneous 96-channel pipetting for convenient processing of both 96-well and 384-well plates.
- Exchangeable dispensing head design covers a wide range of volumes and can accommodate less than 96 tips for maximized flexibility.
- Innovative user interface at the base of the device allows ergonomic use of the epMotion™ 96 Flex in biosafety cabinets.
- Dispensing heads calibrated according to ISO 23783-2 standards, that ensures high pipetting accuracy and precision ensuring reproducibility of your assays.

Specifications
≤ 4.5 % to ≤ 1.0 % / depending on tips and pipette volume | |
Not Autoclavable | |
Polypropylene, Stainless Steel, Polyethylene | |
2-position slider and TipHolder for epMotion™ 96 Flex for use with epT.I.P.S.™ Motion Reloads | |
Air-cushion system | |
≤ 2.5 to 1.5 % depending on tip and pipette volume | |
Polypropylene | |
230 VAC voltage | |
175 W |
Automated Pipetting System | |
96 | |
0.5 to 1.00 ÎĽL | |
Digital | |
Electronic | |
Non-sterile | |
Reload system | |
100/240 V | |
0.729 A |
Product Suggestions
Customers who viewed this item also viewed
Videos
-
epMotion 96 Flex tutorial Automatic tip loading
-
epMotion 96 Flex tutorial smart index 384 Well Plate Platform
-
epMotion 96 Flex tutorial Change of dispensing heads
-
epMotion 96 Flex Flex up your Team
-
epMotion 96 Flex tutorial Automatic tip ejection
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Eppendorf epMotion™ 96 Flex > Channels: 96
Spot an opportunity for improvement?Share a Content Correction