missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Adapters for Rotors- 5430/5430 R centrifuge

For use with rotor F-35-6-30

Brand:  Eppendorf™ 5427746005

101.00 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 15188354

  • 107.00 € / Pack of 2

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • For 1 Eppendorf Tube 5.0mL
  • Available rotor bore sizes: large and small

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Adapter
Small bore hole of 6 x 5mL rotor of Eppendorf Centrifuge 5430/5430 R
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.