missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Adapters for Rotors- 5430/5430 R centrifuge
For use with rotor F-35-6-30
107.00 €
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
15188354
|
Eppendorf™
5427746005 |
Small bore hole of 6 x 5mL rotor of Eppendorf Centrifuge 5430/5430 R |
107.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15178354
|
Eppendorf™
5427747001 |
Large bore hole of 6 x 5mL rotor of Eppendorf Centrifuge 5430/5430 R |
107.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- For 1 Eppendorf Tube 5.0mL
- Available rotor bore sizes: large and small
Specifications
Adapter |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title