missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Heat Sealer Film and Foils
GREENER_CHOICE

Create 100% hermetic seal, eliminate evaporation, and prevent cross-contamination

Brand:  Eppendorf™ 0030127854

145.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 11508663

  • 148.00 € / Pack of 100
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Requires Eppendorf heat sealer (Cat. No. E5390 000 016)
  • Transparent heat sealing film allows sample monitoring in the plates
  • Aluminum coated, heat-sealing foils are available in peel-off or pierceable versions
  • Foil stripper tool removes heat-sealed film and foils completely
  • Certified free of DNA, DNase, RNase, and PCR inhibitors
  • Autoclavable

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Aluminum, Polypropylene
100 Pk.
Automated systems
Yes
Autoclavable
Heat sealing PCR foil for sealing plates, especially recommended for low reaction volumes, best protection against evaporation
PCR Foil
-80°C to +100°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.