Learn More
Eppendorf™ S-4-104 Rotor Adapters
Available in a wide range of configurations to suit a variety of applications.
Brand:  Eppendorf™ 5825740009
 View more versions of this product
Product Code. 13296619
  • 336.34 € / Pack of 2
Explore more special offers
Includes: 2 piece set
Description
Description
  • For use with Centrifuge 5810/5810 R
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rotor Adapter
50 x 1.5/2 mL Tubes
S-4-104 rotor for 5810/5810R centrifuge
2 Pack