missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ S-4-72 Rotor Adapters
Available in a variety of configurations to suite a wide range of applications.
Brand: Eppendorf™ 5804784006
This item is not returnable.
View return policy
Description
- For use with Centrifuge 5804/5804 R and 5810/5810 R

Specifications
Adapter | |
4 x 50 mL Conical Tubes | |
50 mL |
Conical | |
S-4-72 (4 x 250mL) rotor in Model 5804/5810 centrifuges |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Eppendorf™ S-4-72 Rotor Adapters > For 4 x 50mL conical tubes
Spot an opportunity for improvement?Share a Content Correction