Learn More
Eppendorf™ Xplorer™ plus Electronic Pipette, Multi-Channel
GREENER_CHOICE
Xplorer™ plus Multi-Channel Electronic Pipette with intuitive operating system and design for simplicity, precision and reproducibility.
Brand:  Eppendorf™ 4861000821
1239.94 EUR valid until 2025-12-31
Use promo code "24313" to get your promotional price.
 View more versions of this product
Product Code. 12620925
  • 1305.20 € / Each
Explore more special offers
Description
Show More
Specifications
Specifications
Lithium-ion
Green
Pipette Tip (1200 μL)
Xplorer Plus
Electronic Pipette
LCD
1200μL Tip
1 μL
1200μL Tip
Reusable
8
Yes
50 to 1200 μL
Ergonomic
10 Pipetting modes, pipetting history, password protection, a service indicator and the ability to program multiple modes in a sequence for custom applications
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Eppendorf™ Xplorer™ plus Electronic Pipette, Multi-Channel > Multi-Channel Electronic Pipette; Volume: 50 to 1200 μL; Channels: 8; Color: Green Button