Learn More
Eppendorf™ Xplorer™ plus Electronic Pipette, Multi-Channel
GREENER_CHOICE
Xplorer™ plus Multi-Channel Electronic Pipette with intuitive operating system and design for simplicity, precision and reproducibility.
1275.00 € - 1830.00 €
Specifications
Battery Type Lithium-ion
Product Line Xplorer Plus
Type Electronic Pipette
Display LCD
Finger Style Ergonomic
View More Specs
Products 12
Product Code Brand Volume (Metric) Number of Channels Color Price Quantity & Availability  
Product Code Brand Volume (Metric) Number of Channels Color Price Quantity & Availability  
12680915
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
4861000767
0.5 to 10 μL 8 Medium Gray
1275.00 €
Each
12660915
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
4861000775
0.5 to 10 μL 12 Medium Gray
1495.00 €
Each
16262401
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
4861000778
1 to 20 μL 16 Light Pink
1655.00 €
Each
16272401
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
4861000779
1 to 20 μL 24 Light Pink
1830.00 €
Each
12670915
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
4861000783
5 to 100 μL 8 Yellow
1275.00 €
Each
12690915
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
4861000791
5 to 100 μL 12 Yellow
1495.00 €
Each
16282401
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
4861000792
5 to 100 μL 16 Light Yellow
1655.00 €
Each
16292401
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
4861000793
5 to 100 μL 24 Light Yellow
1830.00 €
Each
12600925
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
4861000805
15 to 300 μL 8 Orange
1275.00 €
Each
12610925
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
4861000813
15 to 300 μL 12 Orange
1495.00 €
Each
12620925
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
4861000821
50 to 1200 μL 8 Green
1305.20 €
Each
12630925
GREENER_CHOICE
View accessory products View Documents
Eppendorf™
4861000830
50 to 1200 μL 12 Green
1495.00 €
Each
Description
Description
Xplorer™ plus Multi-Channel Electronic Pipette with intuitive operating system and design for simplicity, precision and reproducibility.
Facilitates performance of additional, intelligent applications including: aspirating, diluting, sequential dispensing and reverse pipetting. Also, helps eliminate delays due to complicated programming and/or inflexible processes. Provides precisely adjustable parameters, maximum reproducible results, fatigue-free work and consistent, full control over the pipetting processes.
  • Multifunctional rocker: ''Up is up and down is down™'' principle
  • Easy selection of all functions with the practical selection dial
  • Operate in the language of your choice
  • Clearly arranged color display; no sub-menus; help function
  • Innovative rocker: When the tip is ejected, the piston automatically returns to zero position
  • ON/OFF switch: Switches the pipetter off during longer periods of inactivity
  • Additional applications include aspirating, diluting, sequential dispensing and reverse pipetting
  • Programming: One-time programming of sequences for easier handling of complex processes
  • Individual fixed volume setting: Desired volume can be saved
  • Password protection: Protects programming and adjustment
  • Individual speed setting: Sets parameters to your individual working speed
  • Pipetter maintenance reminder function, based on time or frequency of use as desired
Reliably reproducible results
  • Tips always rest in the same position
  • Volume and speed precisely and reproducibly adjustable
  • Adjustable to your work environment and requirements
PhysioCare Concept™
  • Reduced weight
  • Improved ergonomics
  • Perfect balance
  • Optimally angled pipetter head
  • Improved view and high resolution display
  • Spring-loaded tip cone provides maximum tightness
Applications
  • Pipetting liquids (manual and automatic)
  • Dispensing (timed and manual)
  • Pipetting and mixing
Specifications
Specifications
Lithium-ion
Electronic Pipette
Ergonomic
Xplorer Plus
LCD
10 Pipetting modes, pipetting history, password protection, a service indicator and the ability to program multiple modes in a sequence for custom applications