Learn More
Eppendorf™ Xplorer™ plus Electronic Pipette, Multi-Channel
GREENER_CHOICE
Xplorer™ plus Multi-Channel Electronic Pipette with intuitive operating system and design for simplicity, precision and reproducibility.
Brand:  Eppendorf™
1420.25 EUR valid until 2025-12-31
Use promo code "24313" to get your promotional price.
 View more versions of this product
Product Code. 12690915
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Show More
Specifications
Specifications
Lithium-ion
Yellow
Pipette Tip (200 μL)
Xplorer Plus
Electronic Pipette
LCD
200μL Tip
10 to 100 μL
200μL Tip
Reusable
12
Yes
5 to 100 μL
Ergonomic
10 Pipetting modes, pipetting history, password protection, a service indicator and the ability to program multiple modes in a sequence for custom applications
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Eppendorf™ Xplorer™ plus Electronic Pipette, Multi-Channel > Multi-Channel Electronic Pipette; Volume: 5 to 100 μL; Channels: 12; Color: Yellow Button