Learn More
Eppendorf epT.I.P.S.™ Motion as Reload System
GREENER_CHOICE
Eppendorf epT.I.P.S. Motion work with the epMotion platform, with a TipHolder accessory the reload trays offer an environmentally friendly option.
Brand:  Eppendorf 0030014502
253.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 30019031
  • 257.00 € / Pack of 2304
Explore more special offers
Description
Description
  • epT.I.P.S. Motion can be used with epMotion™ 96 or integrated into your workstation as a reload system when combined with the tip holder
  • The high-quality reload tips are delivered in a PET thermoformed tray with sealed lid. Upgrading to the autoclavable, aluminum tip holder, which replaces the disposable box, provides waste reduction of up to 40%
  • Eppendorf consumables and thus also epT.I.P.S. Motion pipette tips as Reload System are shipped in packaging consisting of at least 62% recycled cardboard
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10 to 1000 μL
Non-sterile
Not Autoclavable
Hanging Tip Rack
Robotic Tips
Eppendorf epMotion
Polypropylene
Non-filtered
Clear
For Eppendorf epMotion series
Standard, Straight
Single-use
No
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products