Learn More
Eppendorf epT.I.P.S.™ Motion as SafeRack tips
GREENER_CHOICE
The new epMotion systems can reuse tips as e.g. economical choice for multiple wash steps of the same well.
Brand:  Eppendorf 0030014642
138.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 30019047
  • 140.00 € / Pack of 960
Explore more special offers
Description
Description
  • Specially designed for use on the epMotion™ 5070 and 5075 Liquid Handling Workstation. epT.I.P.S. Motion SafeRack pipette tips, with and without filters
  • Developed in torsion-resistant, contamination-free racks that are recommended for the re-use of tips feature of epMotion 5073 systems.
Reuse Tips Feature:
  • The new epMotion systems can reuse tips as e.g. economical choice for multiple wash steps of the same well
  • Pipette tips will be placed back into the position of the box they were removed from
  • Returned tips will be reused within an application for the same source vessel
  • Individual compartments in rack to avoid cross-contamination between used tips
  • Available as standard and PCR clean quality for 50, 300, and 1000 μL
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1.000 μL
Non-sterile
Not Autoclavable
Box
Robotic Tips
Eppendorf epMotion
Polypropylene
Non-filtered
Clear
For Eppendorf epMotion series
Standard, Straight
Single-use
No