Learn More
Eppendorf epT.I.P.S.™ Motion Pipette Tips
GREENER_CHOICE
Eppendorf Totally Integrated Pipetting System for Automation. Works in perfect combination with epMotion platform.
Brand:  Eppendorf 0030015231
255.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 30019049
  • 259.00 € / Pack of 960
Explore more special offers
Description
Description
  • Specially designed for use on the epMotion™ 5070, 5073 and 5075 Liquid Handling Workstation
  • Motion pipette tips, with and without filters, developed in torsion-resistant, clean racks
  • High-precision manufacturing guarantees a perfect fit and extremely accurate dispensing behavior
  • Optimized for simple automatic ejection in the Liquid Handling Workstation collection vessel
  • Can also be integrated into the workstation as a refilling system
  • Packaged in contamination-free racks
  • Available in the following purity grades: standard Eppendorf Quality, PCR clean, Sterile, PCR clean and Sterile
TRUSTED_SUSTAINABILITY
Specifications
Specifications
300 μL
Sterile
Not Autoclavable
Box
Robotic Tips
Eppendorf epMotion
Polypropylene
Filtered
Clear
For Eppendorf epMotion series
Standard, Straight
Single-use
No