Learn More
Eppendorf™ epT.I.P.S.™ Box 2.0 - Pipette Tips
GREENER_CHOICE
Eppendorf epT.I.P.S.™ Box 2.0, available with 'Eppendorf Quality' pipette tips or as an empty box, improves lab efficiency with innovative TwinLid™ technology, enhanced stackability, and superior ergonomics for safe, organized pipetting.
Brand:  Eppendorf™ 0030076176
37.59 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
 View more versions of this product
Product Code. 17382890
  • 39.66 € / Pack of 96
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Contamination-free transfer of trays to stable working box
  • System optimized for use with multichannel pipettes
  • Color-coded trays for simple identification of tips and pipette
  • Tips can be attached to pipette from refill trays
  • Boxes and trays are autoclavable at 121°C for subsequent use
  • epT.I.P.S. Box 2.0 and epT.I.P.S. Set available in Eppendorf Quality
  • ACT™-labeled by non-profit My Green Lab™ to help you make an informed product choice when it comes to sustainability
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 to 1000 μL
Non-sterile
Autoclavable
Eppendorf Quality (Non-sterile)
Box
96 Tips/Cs.
Blue
Air-cushion System
epT.I.P.S.
Non-filtered
Colorless
epTIPS
Eppendorf Pipettes, Non-Eppendorf Pipettes
1 Reusable Box x 96 Tips
Pipette Tips
Single-use
Yes
Polypropylene
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Eppendorf™ epT.I.P.S.™ Box 2.0 - Pipette Tips > Packaging: 1 Reusable Box x 96 Tips, Volume: 50 to 1000 μL, Color Code: Blue, Length: 71 mm