missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Crimper and Decrimper Jaw Sets for Programmable Electronic High-Power Crimp Stations

Optimize your Thermo Scientific™ Programmable Electronic High-Power Crimp Stations with these crimper and decrimper sets.

Brand:  Thermo Scientific™ ECMJ-13FO

344.66 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15277308

  • 346.38 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Optimize your Thermo Scientific™ Programmable Electronic High-Power Crimp Station with these crimper and decrimper sets.

Available in a variety of sizes and formats.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 Ea.
Programmable Electronic High-Power Crimp Station, 11 mm Flip-Off Seal
Industrial Chromatography
Electronic Crimper (Flip-off)
13 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Crimper and Decrimper Jaw Sets for Programmable Electronic High-Power Crimp Stations >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.