missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) fluoride, anhydrous, REacton™, 99.99% (REO), Thermo Scientific Chemicals

162.00 € - 552.00 €

Chemical Identifiers

CAS 13760-83-3
Molecular Formula ErF3
Molecular Weight (g/mol) 224.25
MDL Number MFCD00016074
InChI Key QGJSAGBHFTXOTM-UHFFFAOYSA-K
IUPAC Name erbium(3+) trifluoride
SMILES [F-].[F-].[F-].[Er+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11371059
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011312.09
10 g
162.00 €
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11361059
View Documents Promotion Details
Thermo Scientific Alfa Aesar
011312.18
50 g
552.00 €
50g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Erbium(III) fluoride is an important colorant in glass manufacturing and porcelain enamel glazes. It is used to produce erbium metal. It is used as dopant in making optical products. It is highly employed as erbium doped fluoride fibers in optical communications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13760-83-3
224.25
QGJSAGBHFTXOTM-UHFFFAOYSA-K
[F-].[F-].[F-].[Er+3]
ErF3
MFCD00016074
erbium(3+) trifluoride

Specifications

1350°C
Granules, Powder
MFCD00016074
QGJSAGBHFTXOTM-UHFFFAOYSA-K
erbium(3+) trifluoride
224.28
Reacton™
Hygroscopic
(REO)
13760-83-3
ErF3
Insoluble in water.
[F-].[F-].[F-].[Er+3]
224.25
99.99%
Odorless
6.4 g/mL
Erbium(III) fluoride, Anhydrous
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 237-356-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only