missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) nitrate hydrate, 99.9% (REO), Thermo Scientific Chemicals

116.58 € - 368.84 €

Chemical Identifiers

CAS 10031-51-3
Molecular Formula ErH10N3O14
Molecular Weight (g/mol) 443.35
MDL Number MFCD00149691
InChI Key LWHHUEHWVBVASY-UHFFFAOYSA-N
Synonym erbium iii nitrate pentahydrate, erbium trinitrate pentahydrate, erbium nitrate pentahydrate, acmc-20al2v, er.3no3.5h2o, erbium nitrate-water 1/3/5, erbium iii nitrate pentahydrate, reacton, erbium nitrate, pentahydrate,
PubChem CID 53249207
SMILES O.O.O.O.O.[Er+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11317657
SDS View Documents
Thermo Scientific Alfa Aesar
012920.14
25 g
116.58 €
25g
Please to purchase this item. Need a web account? Register with us today!
11307657
SDS View Documents
Thermo Scientific Alfa Aesar
012920.22
100 g
368.84 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Used as laboratory chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10031-51-3
443.35
LWHHUEHWVBVASY-UHFFFAOYSA-N
53249207
ErH10N3O14
MFCD00149691
erbium iii nitrate pentahydrate, erbium trinitrate pentahydrate, erbium nitrate pentahydrate, acmc-20al2v, er.3no3.5h2o, erbium nitrate-water 1/3/5, erbium iii nitrate pentahydrate, reacton, erbium nitrate, pentahydrate,
O.O.O.O.O.[Er+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

130°C (in -4H2O)
Crystalline
MFCD00149691
erbium iii nitrate pentahydrate, erbium trinitrate pentahydrate, erbium nitrate pentahydrate, acmc-20al2v, er.3no3.5h2o, erbium nitrate-water 1/3/5, erbium iii nitrate pentahydrate, reacton, erbium nitrate, pentahydrate,
LWHHUEHWVBVASY-UHFFFAOYSA-N
erbium(3+) pentahydrate trinitrate
53249207
99.9%
(REO)
10031-51-3
ErH10N3O14
UN1477
Typically insoluble in water.
O.O.O.O.O.[Er+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
443.35
353.27 (Anhydrous)
Hygroscopic
Erbium(III) nitrate hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H272-H315-H319-H335

missing translation for 'dotInformation' : Hazard Class: 5.1; Packaging Group: II

missing translation for 'einecsNumber' : 233-436-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.