Learn More
Ertugliflozin, MedChemExpress
MedChemExpress Ertugliflozin (PF-04971729) is a potent, selective and orally active inhibitor of the sodium-dependent glucose cotransporter 2 (SGLT2), with an IC50 of 0.877 nM for h-SGLT2. Has the potential for the treatment of type 2 diabetes mellitus.
70.20 € - 167.40 €
Specifications
Chemical Name or Material | Ertugliflozin |
---|---|
CAS | 1210344-57-2 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C22H25ClO7 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16496538
![]() |
Medchem Express
HY-15461 |
10 mg |
97.20 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16429127
![]() |
Medchem Express
HY-15461 |
5 mg |
70.20 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16422637
![]() |
Medchem Express
HY-15461 |
50 mg |
167.40 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ertugliflozin (PF-04971729) is a potent, selective and orally active inhibitor of the sodium-dependent glucose cotransporter 2 (SGLT2), with an IC50 of 0.877 nM for h-SGLT2. Has the potential for the treatment of type 2 diabetes mellitus.Specifications
Ertugliflozin | |
White | |
C22H25ClO7 | |
DMSO : 250 mg/mL (572.24 mM; Need ultrasonic) | |
436.88 | |
98.82% | |
Solid |
1210344-57-2 | |
Research | |
PF-04971729 | |
ClC1=CC=C([C@]23O[C@@](CO)(CO3)[C@@H](O)[C@H](O)[C@H]2O)C=C1CC4=CC=C(OCC)C=C4 | |
436.88 | |
Research | |
Metabolism-sugar/lipid metabolism |
Safety and Handling
H315∣H317∣H318∣H334∣H335∣H341∣H361∣H370∣H413
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.