missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Estradiol 3-sulfamate, MedChemExpress
MedChemExpress Estradiol 3-sulfamate (BLE 00084; E2MATE; ES-J 995) is a potent, long-acting, and orally active steroid sulfatase inhibitor; inhibits estrone sulfatase with an IC50 of 251 nM and a Ki of 133 nM.
318.60 € - 1912.95 €
Specifications
Chemical Name or Material | Estradiol 3-sulfamate |
---|---|
CAS | 172377-52-5 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C18H25NO4S |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16482368
|
Medchem Express
HY-U00112 |
5 mg |
637.20 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16480686
|
Medchem Express
HY-U00112 |
20 mg |
1912.95 €
20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16480169
|
Medchem Express
HY-U00112 |
10 mg |
1084.05 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16423159
|
Medchem Express
HY-U00112 |
1 mg |
318.60 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Estradiol 3-sulfamate (BLE 00084Specifications
Estradiol 3-sulfamate | |
White | |
C18H25NO4S | |
DMSO : 62.5 mg/mL (177.83 mM; Need ultrasonic) | |
351.46 | |
99.5% | |
Solid |
172377-52-5 | |
Research | |
BLE 00084 E2MATE ES-J 995 | |
C[C@@]1([C@H]2O)[C@](CC2)([H])[C@@](CCC3=C4C=CC(OS(N)(=O)=O)=C3)([H])[C@]4([H])CC1 | |
351.46 | |
Research | |
Cancer-programmed cell death |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only