Learn More
Estramustine phosphate sodium, MedChemExpress
MedChemExpress Estramustine phosphate sodium, an estradiol analog, is an orally active antimicrotubule chemotherapy agent. Estramustine phosphate sodium depolymerises microtubules by binding to microtubule associated proteins (MAPs) and/or to tubulin. Estramustine phosphate sodium can interfere mitosis, trigger cell death and induce apoptosis, which can be used for the research of cancer like prostate cancer.
Brand: Medchem Express HY-13627
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Estramustine phosphate sodium, an estradiol analog, is an orally active antimicrotubule chemotherapy agent. Estramustine phosphate sodium depolymerises microtubules by binding to microtubule associated proteins (MAPs) and/or to tubulin. Estramustine phosphate sodium can interfere mitosis, trigger cell death and induce apoptosis, which can be used for the research of cancer like prostate cancer.Spécification
Estramustine phosphate sodium | |
White | |
C23H30Cl2NNa2O6P | |
H2O : 62.5 mg/mL (110.75 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (8.86 mM; Need ultrasonic) | |
564.35 | |
96.79% | |
Solid |
52205-73-9 | |
Research | |
50 mg | |
O=C(N(CCCl)CCCl)OC1=CC2=C(C=C1)[C@@]3([H])CC[C@@]4(C)[C@](CC[C@@H]4OP(O[Na])(O[Na])=O)([H])[C@]3([H])CC2 | |
564.35 | |
Research | |
Cancer-programmed cell death |
Research purposes only